Ethyl 2-amino-2-methylpropanoate 95%
Lieferant: Apollo Scientific
OR939867-1G
APOSOR939867-1GEA
433
EUR
APOSOR939867-1G
Ethyl 2-amino-2-methylpropanoate 95%
Ethyl 2-amino-2-methylpropanoate
Formel:
C₆H₁₃NO₂ Molecular Weight: 131,173 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02663028 CAS-Nummer: 1113-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...