3-(Carboxymethyl)benzoic acid 95%

Lieferant: Apollo Scientific

OR939956-1G OR939956-5G
APOSOR939956-1GEA 0 EUR
APOSOR939956-1G APOSOR939956-5G
3-(Carboxymethyl)benzoic acid 95%
3-(Carboxymethyl)benzoic acid
Formel: C₉H₈O₄
Molecular Weight: 180.157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15523567
CAS-Nummer: 2084-13-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR