3-(Carboxymethyl)benzoic acid 95%
Lieferant: Apollo Scientific
OR939956-1G
OR939956-5G
APOSOR939956-1GEA
0
EUR
APOSOR939956-1G
APOSOR939956-5G
3-(Carboxymethyl)benzoic acid 95%
3-(Carboxymethyl)benzoic acid
Formel:
C₉H₈O₄ Molecular Weight: 180.157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15523567 CAS-Nummer: 2084-13-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...