sec-Butyl butyrate 95%

Lieferant: Apollo Scientific

Synonyme: Butyric Acid sec-Butyl Ester

OR939965-10G OR939965-1G OR939965-5G
APOSOR939965-10GEA 328 EUR
APOSOR939965-10G APOSOR939965-1G APOSOR939965-5G
sec-Butyl butyrate 95%
sec-Butyl butyrate
Formel: C₈H₁₆O₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 819-97-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR