5-(Chloromethyl)furan-2-carbaldehyde 95%

Lieferant: Apollo Scientific

OR939970-1G
APOSOR939970-1GEA 397 EUR
APOSOR939970-1G
5-(Chloromethyl)furan-2-carbaldehyde 95%
5-(Chloromethyl)furan-2-carbaldehyde
Formel: C₆H₅ClO₂
Molecular Weight: 144,556 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02181084
CAS-Nummer: 1623-88-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR