Sorbic chloride 80%

Lieferant: Apollo Scientific

OR939977-1G OR939977-5G
APOSOR939977-1GEA 0 EUR
APOSOR939977-1G APOSOR939977-5G
Sorbic chloride 80%
Sorbic chloride
Formel: C₆H₇ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD00045227
CAS-Nummer: 2614-88-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR