6-Chloroimidazo[1,2-b]pyridazine-3-carbonitrile 95%

Lieferant: Apollo Scientific

OR940590-1G OR940590-500MG OR940590-250MG
APOSOR940590-1GEA 313 EUR
APOSOR940590-1G APOSOR940590-500MG APOSOR940590-250MG
6-Chloroimidazo[1,2-b]pyridazine-3-carbonitrile 95%
6-Chloroimidazo[1,2-b]pyridazine-3-carbonitrile
Formel: C₇H₃ClN₄
Lagertemperatur: Raumtemperatur
MDL: MFCD11044763
CAS-Nummer: 123531-54-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR