3-Ethoxy-4-methoxybenzonitrile
Lieferant: Apollo Scientific
OR940598-100G
APOSOR940598-100GEA
85.7
EUR
APOSOR940598-100G
3-Ethoxy-4-methoxybenzonitrile
3-Ethoxy-4-methoxybenzonitrile
Formel:
C₁₀H₁₁NO₂ Molecular Weight: 177,20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02256160 CAS-Nummer: 60758-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...