Gimeracil 95%

Lieferant: Apollo Scientific

Synonyme: 5-Chloro-4-hydroxypyridin-2(1H)-one, 5-Chloro-4-hydroxy-2(1H)-pyridinone, 5-Chloro-2,4-pyridinediol, 5-Chloro-2,4-dihydroxypyridine

OR940679-100G
APOSOR940679-100GEA 126 EUR
APOSOR940679-100G
Gimeracil 95%
Gimeracil
Formel: C₅H₄ClNO₂
Molecular Weight: 145.54 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458352
CAS-Nummer: 103766-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR