4-Brom-2-(pyrrolidin-1-ylsulfonyl)anisol 95%

Lieferant: Apollo Scientific

Synonyme: 1-((5-Bromo-2-methoxyphenyl)sulphonyl)pyrrolidine

OR9407-1G
APOSOR9407-1GEA 154 EUR
APOSOR9407-1G
4-Brom-2-(pyrrolidin-1-ylsulfonyl)anisol 95%
4-Brom-2-(pyrrolidin-1-ylsulfonyl)anisol
Formel: C₁₁H₁₄BrNO₃S
Molecular Weight: 320,2000 g/mol
Schmelzpunkt: 116…120 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04091911
CAS-Nummer: 691381-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR