5-Iodo-2-methyl-1H-benzo[d]imidazole ≥98%

Lieferant: Apollo Scientific

OR940767-1G
APOSOR940767-1GEA 0 EUR
APOSOR940767-1G
5-Iodo-2-methyl-1H-benzo[d]imidazole ≥98%
5-Iodo-2-methyl-1H-benzo[d]imidazole
Formel: C₈H₇IN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00457529
CAS-Nummer: 2818-70-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR