2-Chloro-N-(2-oxotetrahydrothiophen-3-yl)acetamide 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-N-(2-oxothiolan-3-yl)acetamide

OR940774-1G
APOSOR940774-1GEA 386 EUR
APOSOR940774-1G
2-Chloro-N-(2-oxotetrahydrothiophen-3-yl)acetamide 95%
2-Chloro-N-(2-oxotetrahydrothiophen-3-yl)acetamide
Formel: C₆H₈ClNO₂S
Molecular Weight: 193,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07393747
CAS-Nummer: 84611-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR