Ethylen Glykol Mono-T-Butyl Ether 95%

Lieferant: Apollo Scientific

OR940810-100G OR940810-1KG OR940810-500G
APOSOR940810-100GEA 125 EUR
APOSOR940810-100G APOSOR940810-1KG APOSOR940810-500G
Ethylen Glykol Mono-T-Butyl Ether 95%
Ethylene glycol mono-tert-butyl ether
Formel: C₆H₁₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00059004
CAS-Nummer: 7580-85-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR