5-Oxa-8-azaspiro[3.5]nonane

Lieferant: Apollo Scientific

OR940852-1G
APOSOR940852-1GEA 0 EUR
APOSOR940852-1G
5-Oxa-8-azaspiro[3.5]nonane
5-Oxa-8-azaspiro[3.5]nonane
Formel: C₇H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD18382670
CAS-Nummer: 220291-93-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR