trans-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]cyclobutanol

Lieferant: Apollo Scientific

OR940901-1G OR940901-250MG
APOSOR940901-1GEA 867 EUR
APOSOR940901-1G APOSOR940901-250MG
trans-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]cyclobutanol
trans-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]cyclobutanol
Formel: C₁₀H₂₂O₂Si
Lagertemperatur: Raumtemperatur
MDL: MFCD23105931
CAS-Nummer: 1408075-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR