Ethyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate 95%

Lieferant: Apollo Scientific

OR94092-250MG
APOSOR94092-250MGEA 84.4 EUR
APOSOR94092-250MG
Ethyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate 95%
Ethyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate
Formel: C₁₉H₃₀O₃
Molecular Weight: 306,44 g/mol
MDL: MFCD00210464
CAS-Nummer: 36294-24-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR