1-tert-Butyl-2-nitrobenzol 95%

Lieferant: Apollo Scientific

Synonyme: 2-tert-Butylnitrobenzol

OR941016-1G
APOSOR941016-1GEA 21.9 EUR
APOSOR941016-1G
1-tert-Butyl-2-nitrobenzol 95%
1-tert-Butyl-2-nitrobenzol
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082705
CAS-Nummer: 1886-57-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR