ɑ,ɑ'-Dihydroxy-1,3-diisopropylbenzene 95%
Lieferant: Apollo Scientific
Synonyme:
1,3-Bis(α-hydroxyisopropyl)benzene, α,α'-Dihydroxy-1,3-diisopropylbenzene, α,α,α',α'-Tetramethyl-1,3-benzenedimethanol, ɑ,ɑ,ɑ',ɑ'-Tetramethyl-1,3-benzenedimethanol, 1,3-Bis(ɑ-hydroxyisopropyl)benzene, 2,2'-(m-Phenylene)di-2-propanol
OR941073-25G
APOSOR941073-25GEA
154
EUR
APOSOR941073-25G
ɑ,ɑ'-Dihydroxy-1,3-diisopropylbenzene 95%
alpha,alpha'-Dihydroxy-1,3-diisopropylbenzene
Formel:
C₁₂H₁₈O₂ Molecular Weight: 194,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059139 CAS-Nummer: 1999-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...