2-Methyl-3-piperidin-1-yl-propan-1-ol 95%
Lieferant: Apollo Scientific
OR941090-1G
APOSOR941090-1GEA
522
EUR
APOSOR941090-1G
2-Methyl-3-piperidin-1-yl-propan-1-ol 95%
2-Methyl-3-piperidin-1-yl-propan-1-ol
Formel:
C₉H₁₉NO Molecular Weight: 157,253 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08061223 CAS-Nummer: 62101-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...