N,5-Dimethylpyridin-2-amine 95%
Lieferant: Apollo Scientific
OR941253-1G
OR941253-250MG
APOSOR941253-1GEA
0
EUR
APOSOR941253-1G
APOSOR941253-250MG
N,5-Dimethylpyridin-2-amine 95%
N,5-Dimethylpyridin-2-amine
Formel:
C₇H₁₀N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06411230 CAS-Nummer: 45715-13-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...