Methyl-2-methoxy-6-methylbenzoate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-methoxy-6-methylbenzoate

OR941298-1G
APOSOR941298-1GEA 95.7 EUR
APOSOR941298-1G
Methyl-2-methoxy-6-methylbenzoate 95%
Methyl-2-methoxy-6-methylbenzoate
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08235005
CAS-Nummer: 79383-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR