3,4-Dihydro-2-ethoxy-2H-pyran 95%
Lieferant: Apollo Scientific
OR941406-5G
APOSOR941406-5GEA
82.8
EUR
APOSOR941406-5G
3,4-Dihydro-2-ethoxy-2H-pyran 95%
3,4-Dihydro-2-ethoxy-2H-pyran
Formel:
C₇H₁₂O₂ Molecular Weight: 128,17 g/mol Siedepunkt: 132…133 °C Dichte: 0,966 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
CAS-Nummer:
103-75-3 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...