4-Brom-2-(morpholin-4-ylsulfonyl)anisol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-((5-Bromo-2-methoxyphenyl)sulphonyl)morpholine

OR9415-1G
APOSOR9415-1GEA 154 EUR
APOSOR9415-1G
4-Brom-2-(morpholin-4-ylsulfonyl)anisol 98%
4-Brom-2-(morpholin-4-ylsulfonyl)anisol
Formel: C₁₁H₁₄BrNO₄S
Molecular Weight: 336,2000 g/mol
Schmelzpunkt: 151…155 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02043681
CAS-Nummer: 325809-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR