5-Chloro-2-hydroxy-6-methylpyridine-3-carboxylic acid 95%

Lieferant: Apollo Scientific

OR941579-1G
APOSOR941579-1GEA 0 EUR
APOSOR941579-1G
5-Chloro-2-hydroxy-6-methylpyridine-3-carboxylic acid 95%
5-Chloro-2-hydroxy-6-methylpyridine-3-carboxylic acid
Formel: C₇H₆ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11044289
CAS-Nummer: 117449-75-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR