1,2,3,4-Tetrahydroisoquinolin-5-ol 95%

Lieferant: Apollo Scientific

OR941622-1G OR941622-250MG
APOSOR941622-1GEA 520 EUR
APOSOR941622-1G APOSOR941622-250MG
1,2,3,4-Tetrahydroisoquinolin-5-ol 95%
1,2,3,4-Tetrahydroisoquinolin-5-ol
Formel: C₉H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD06656937
CAS-Nummer: 102877-50-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR