Butyl 2-oxoacetate 95%
Lieferant: Apollo Scientific
OR941629-1G
APOSOR941629-1GEA
0
EUR
APOSOR941629-1G
Butyl 2-oxoacetate 95%
Butyl 2-oxoacetate
Formel:
C₆H₁₀O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043908 CAS-Nummer: 6295-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...