(5-Methoxy-2-nitrophenyl)methanol 98%

Lieferant: Apollo Scientific

OR941669-1G
APOSOR941669-1GEA 319 EUR
APOSOR941669-1G
(5-Methoxy-2-nitrophenyl)methanol 98%
(5-Methoxy-2-nitrophenyl)methanol
Formel: C₈H₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09261036
CAS-Nummer: 879-55-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR