4,5-Dibrom-1-tritylimidazol 98%

Lieferant: Apollo Scientific

Synonyme: 4,5-Dibromo-1-triphenylmethyl-1H-imidazole

OR9417-1G
APOSOR9417-1GEA 176 EUR
APOSOR9417-1G
4,5-Dibrom-1-tritylimidazol 98%
4,5-Dibrom-1-tritylimidazol
Formel: C₂₂H₁₆Br₂N₂
Schmelzpunkt: 223…225 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08056373
CAS-Nummer: 112517-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR