N,N,2,2-Tetramethyl-1,3-propandiamin 95%

Lieferant: Apollo Scientific

Synonyme: N,N-Dimethylneopentanediamine, N1,N1,2,2-Tetramethylpropane-1,3-diamine

OR941777-100G
APOSOR941777-100GEA 89.9 EUR
APOSOR941777-100G
N,N,2,2-Tetramethyl-1,3-propandiamin 95%
N,N,2,2-Tetramethyl-1,3-propandiamin
Formel: C₇H₁₈N₂
Molecular Weight: 130.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00044018
CAS-Nummer: 53369-71-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR