2-Chloro-3-methoxy-6-methylpyridine 95%

Lieferant: Apollo Scientific

OR941863-1G OR941863-250MG
APOSOR941863-1GEA 212 EUR
APOSOR941863-1G APOSOR941863-250MG
2-Chloro-3-methoxy-6-methylpyridine 95%
2-Chloro-3-methoxy-6-methylpyridine
Formel: C₇H₈ClNO
Molecular Weight: 157.598 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09260845
CAS-Nummer: 204378-42-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR