3-(N-Ethylaminocarbonyl)-5-nitrobenzeneboronic acid 98%
Lieferant: Apollo Scientific
Synonyme:
3-(Ethylcarbamoyl)-5-nitrophenylboronic acid
OR9419-1G
APOSOR9419-1GEA
176
EUR
APOSOR9419-1G
3-(N-Ethylaminocarbonyl)-5-nitrobenzeneboronic acid 98%
3-(N-Ethylaminocarbonyl)-5-nitrobenzeneboronic acid
Formel:
C₉H₁₁BN₂O₅ Molecular Weight: 238,005 g/mol Schmelzpunkt: 187…191 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07783872 CAS-Nummer: 871332-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...