Methyl 4-formyl-3-methoxybenzoate 95%

Lieferant: Apollo Scientific

OR941918-1G
APOSOR941918-1GEA 125 EUR
APOSOR941918-1G
Methyl 4-formyl-3-methoxybenzoate 95%
Methyl 4-formyl-3-methoxybenzoate
Formel: C₁₀H₁₀O₄
Molecular Weight: 194,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17169951
CAS-Nummer: 74733-24-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR