γ-chlorocrotonic acid 90%

Lieferant: Apollo Scientific

OR941955-1G
APOSOR941955-1GEA 478 EUR
APOSOR941955-1G
γ-chlorocrotonic acid 90%
γ-chlorocrotonic acid
Formel: C₄H₅ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19228094
CAS-Nummer: 16197-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR