4-Allyloxybenzoic acid 95%

Lieferant: Apollo Scientific

OR941983-1G
APOSOR941983-1GEA 68.3 EUR
APOSOR941983-1G
4-Allyloxybenzoic acid 95%
4-Allyloxybenzoic acid
Formel: C₁₀H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00526542
CAS-Nummer: 27914-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR