3-(Benzylaminocarbonyl)-5-nitrobenzeneboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: 3-(Benzylcarbamoyl)-5-nitrobenzeneboronic acid

OR9420-1G
APOSOR9420-1GEA 176 EUR
APOSOR9420-1G
3-(Benzylaminocarbonyl)-5-nitrobenzeneboronic acid 98%
3-(Benzylaminocarbonyl)-5-nitrobenzeneboronic acid
Formel: C₁₄H₁₃BN₂O₅
Molecular Weight: 300,074 g/mol
Schmelzpunkt: 246…248 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07783882
CAS-Nummer: 871332-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR