1-(1H-Tetrazol-5-yl)piperidine

Lieferant: Apollo Scientific
Achtung

OR9421-100MG
APOSOR9421-100MGEA 29.2 EUR
APOSOR9421-100MG
1-(1H-Tetrazol-5-yl)piperidine
1-(1H-Tetrazol-5-yl)piperidine
Formel: C₆H₁₁N₅
Lagertemperatur: Raumtemperatur
MDL: MFCD01861992
CAS-Nummer: 6280-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR