2-Methoxy-6-methylbenzamide 95%
Lieferant: Apollo Scientific
OR942134-1G
APOSOR942134-1GEA
647
EUR
APOSOR942134-1G
2-Methoxy-6-methylbenzamide 95%
2-Methoxy-6-methylbenzamide
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,189 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13187522 CAS-Nummer: 139583-90-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...