6-(Methoxymethyl)-2-(methylthio)pyrimidin-4-ol

Lieferant: Apollo Scientific

Synonyme: 4-Hydroxy-6-(methoxymethyl)-2-(methylthio)pyrimidine

OR9422-1G
APOSOR9422-1GEA 183 EUR
APOSOR9422-1G
6-(Methoxymethyl)-2-(methylthio)pyrimidin-4-ol
6-(Methoxymethyl)-2-(methylthio)pyrimidin-4-ol
Formel: C₇H₁₀N₂O₂S
Molecular Weight: 186.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052618
CAS-Nummer: 68087-13-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR