2,3-Piperazindion 95%

Lieferant: Apollo Scientific
Achtung

OR942215-1G
APOSOR942215-1GEA 212 EUR
APOSOR942215-1G
2,3-Piperazindion 95%
2,3-Piperazindion
Formel: C₄H₆N₂O₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 13092-86-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR