3-(Piperidin-1-ylmethyl)benzoic acid 97%

Lieferant: Apollo Scientific

OR942220-1G
APOSOR942220-1GEA 314 EUR
APOSOR942220-1G
3-(Piperidin-1-ylmethyl)benzoic acid 97%
3-(Piperidin-1-ylmethyl)benzoic acid
Formel: C₁₃H₁₇NO₂
Molecular Weight: 219.29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05841589
CAS-Nummer: 158861-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR