3-(Aminocarbonyl)-5-nitrobenzeneboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Carbamoyl-5-nitrobenzeneboronic acid, (3-Carbamoyl-5-nitrophenyl)boronic acid

OR9423-250MG
APOSOR9423-250MGEA 46.4 EUR
APOSOR9423-250MG
3-(Aminocarbonyl)-5-nitrobenzeneboronic acid 98%
3-(Aminocarbonyl)-5-nitrobenzeneboronic acid
Formel: C₇H₇BN₂O₅
Molecular Weight: 209,952 g/mol
Schmelzpunkt: 269…271 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07783870
CAS-Nummer: 102170-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR