Methyl 3-(4-formylphenyl)acrylate 95%

Lieferant: Apollo Scientific
Achtung

OR942301-1G OR942301-5G
APOSOR942301-1GEA 42.2 EUR
APOSOR942301-1G APOSOR942301-5G
Methyl 3-(4-formylphenyl)acrylate 95%
Methyl 3-(4-formylphenyl)acrylate
Formel: C₁₁H₁₀O₃
Schmelzpunkt: 86…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00157179
CAS-Nummer: 58045-41-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR