2,3-Dihydrobenzo[b]furan-7-methanol 95%

Lieferant: Apollo Scientific

OR942316-100MG
APOSOR942316-100MGEA 78.3 EUR
APOSOR942316-100MG
2,3-Dihydrobenzo[b]furan-7-methanol 95%
2,3-Dihydrobenzo[b]furan-7-methanol
Formel: C₉H₁₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06411535
CAS-Nummer: 151155-53-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR