3-Cyano-5-methylbenzoic acid 95%
Lieferant: Apollo Scientific
OR942318-1G
APOSOR942318-1GEA
50.9
EUR
APOSOR942318-1G
3-Cyano-5-methylbenzoic acid 95%
3-Cyano-5-methylbenzoic acid
Formel:
C₉H₇NO₂ Molecular Weight: 161,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11007822 CAS-Nummer: 78621-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...