5-Methoxybenzo[b]thiophene-2-carbaldehyde 95%

Lieferant: Apollo Scientific

OR942338-1G
APOSOR942338-1GEA 248 EUR
APOSOR942338-1G
5-Methoxybenzo[b]thiophene-2-carbaldehyde 95%
5-Methoxybenzo[b]thiophene-2-carbaldehyde
Formel: C₁₀H₈O₂S
Molecular Weight: 192,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11101036
CAS-Nummer: 622864-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR