3-(Isopropylcarbamoyl)-5-nitrobenzeneboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: 3-(Isopropylcarbamoyl)-5-nitrobenzeneboronic acid

OR9424-1G
APOSOR9424-1GEA 176 EUR
APOSOR9424-1G
3-(Isopropylcarbamoyl)-5-nitrobenzeneboronic acid 98%
3-(Isopropylcarbamoyl)-5-nitrobenzeneboronic acid
Formel: C₁₀H₁₃BN₂O₅
Molecular Weight: 252,032 g/mol
Schmelzpunkt: 198…201 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07783875
CAS-Nummer: 871332-83-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR