2-Bromo-3-hydroxybenzonitrile 95%

Lieferant: Apollo Scientific

OR942456-1G OR942456-250MG
APOSOR942456-1GEA 223 EUR
APOSOR942456-1G APOSOR942456-250MG
2-Bromo-3-hydroxybenzonitrile 95%
2-Bromo-3-hydroxybenzonitrile
Formel: C₇H₄BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09266204
CAS-Nummer: 693232-06-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR