3-[(N-Methoxy-N-methylamino)carbonyl]-5-nitrobenzeneboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: 3-[Methoxy(methyl)carbamoyl]-5-nitrophenylboronic acid

OR9425-1G
APOSOR9425-1GEA 190 EUR
APOSOR9425-1G
3-[(N-Methoxy-N-methylamino)carbonyl]-5-nitrobenzeneboronic acid 98%
3-[(N-Methoxy-N-methylamino)carbonyl]-5-nitrobenzeneboronic acid
Formel: C₉H₁₁BN₂O₆
Molecular Weight: 254,004 g/mol
Schmelzpunkt: 156…160 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07783876
CAS-Nummer: 871332-84-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR