Imidazo[1,2-a]pyridine-3-carbonitrile 95%
Lieferant: Apollo Scientific
OR942541-1G
APOSOR942541-1GEA
122
EUR
APOSOR942541-1G
Imidazo[1,2-a]pyridine-3-carbonitrile 95%
Imidazo[1,2-a]pyridine-3-carbonitrile
Formel:
C₈H₅N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01720448 CAS-Nummer: 6200-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...