2-Cyanomethyl benzoic acid ethyl ester 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-(cyanomethyl)benzoate
OR942594-1G
APOSOR942594-1GEA
77
EUR
APOSOR942594-1G
2-Cyanomethyl benzoic acid ethyl ester 95%
2-Cyanomethyl benzoic acid ethyl ester
Formel:
C₁₁H₁₁NO₂ Molecular Weight: 189,211 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00062509 CAS-Nummer: 19821-21-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...